Learn More
Analytical Sales & Services PEEK Microtight Tubing Sleeve for 175-215µm OD Tubing
Catalog No. 5000805061
Encompass
PEEK Microtight Tubing Sleeve\n\n• Length: 1.6″\n• Inner Diameter: 230µm (0.009″), for 175-215µm OD Tubing\n• Outer Diameter: 1/8″\n• Color: Natural
Catalog No. 50-008-05061 Supplier Analytical Sales & Services Supplier No. 55182X
PEEK Microtight Tubing Sleeve\n\n• Length: 1.6″\n• Inner Diameter: 230µm (0.009″), for 175-215µm OD Tubing\n• Outer Diameter: 1/8″\n• Color: Natural