Learn More
Analytical Sales & Services PEEK Microtight Tubing Sleeve for 340-380µm OD Tubing
Catalog No. 5000805062
Encompass
PEEK Microtight Tubing Sleeve\n\n• Length: 1.6″\n• Inner Diameter: 395µm (0.0155″), for 340-380µm OD Tubing\n• Outer Diameter: 1/8″\n• Color: Green
Catalog No. 50-008-05062 Supplier Analytical Sales & Services Supplier No. 55185X
PEEK Microtight Tubing Sleeve\n\n• Length: 1.6″\n• Inner Diameter: 395µm (0.0155″), for 340-380µm OD Tubing\n• Outer Diameter: 1/8″\n• Color: Green