Learn More
Analytical Sales & Services PEEK Microtight Union Assembly 6-32 To 6-32
Catalog No. 50450875
Encompass
This item has a minimum order quantity of 2 per supplier requirements.PEEK Microtight Union Assembly 6-32 to 6-32 Includes; (2) 55126S Fittings and (1) 59553 Gauge Plug No Sleeves Required for the Connectors
Catalog No. 50-450-875 Supplier Analytical Sales & Services Supplier No. 59771
This item has a minimum order quantity of 2 per supplier requirements.PEEK Microtight Union Assembly 6-32 to 6-32 Includes; (2) 55126S Fittings and (1) 59553 Gauge Plug No Sleeves Required for the Connectors