Learn More
Alzet PEEK Tubing for Use with 200 µl ALZET Osmotic Pumps, Ideal in MRI Studies as a Non-metallic Replacement for the ALZET Flow Moderator
SDP
Catalog No. NC1571629
Encompass
ALZETTM PEEK Tubing, pre-cut and sterilized, is a non-metallic replacement for the flow moderator of an ALZET Osmotic Pump for use in studies involving MRI. PEEK has excellent chemical compatibility and is widely used in medical applications. This item, PEEK Tube for 200 µl ALZET Pumps, is the optimal length and diameter for ALZET Pump Models 2001D, 2001, 2002, 2004, 2006. Instructions for use can be found at https://www.alzet.com/guide-to-use/mri/
Catalog No. NC1571629 Supplier Alzet Supplier No. 0002496
ALZETTM PEEK Tubing, pre-cut and sterilized, is a non-metallic replacement for the flow moderator of an ALZET Osmotic Pump for use in studies involving MRI. PEEK has excellent chemical compatibility and is widely used in medical applications. This item, PEEK Tube for 200 µl ALZET Pumps, is the optimal length and diameter for ALZET Pump Models 2001D, 2001, 2002, 2004, 2006. Instructions for use can be found at https://www.alzet.com/guide-to-use/mri/