Learn More
IDEX Health & Science Upchurch LiteTouch PEEK Nut ,for 1/16" OD Single
Catalog No. NC1793763
Encompass_Preferred
Our LiteTouch PEEK Nut is designed to help prevent the twisting of polymer tubing by gripping tubing at two compression points. It's unique design allows it to be used on hard tubing like stainless steel or PEEK tubing preventing the collapse of tubing ID. It is for 1/16"OD tubing.
Catalog No. NC1793763 Supplier IDEX Health & Science Upchurch Supplier No. LT110
Our LiteTouch PEEK Nut is designed to help prevent the twisting of polymer tubing by gripping tubing at two compression points. It's unique design allows it to be used on hard tubing like stainless steel or PEEK tubing preventing the collapse of tubing ID. It is for 1/16"OD tubing.