Learn More
Analytical Sales & Services PEEK Flangeless Male Nut, Biocompatible, Reusable, 1/8 in Tubing
Catalog No. NC1374215
Encompass
Introducing the PEEK Flangeless Nut designed for 1/8 in OD tubing. This product ensures robust connections in chromatography systems due to its biocompatible and chemical resistant construction. Its flangeless design simplifies installation while maintaining reusable features and excellent pressure-holding capabilities up to 500 psi, making it ideal for sensitive analytical processes. Trust this superior construction to enhance performance and reliability in your lab applications.
Catalog No. NC1374215 Supplier Analytical Sales & Services Supplier No. 59330X
Introducing the PEEK Flangeless Nut designed for 1/8 in OD tubing. This product ensures robust connections in chromatography systems due to its biocompatible and chemical resistant construction. Its flangeless design simplifies installation while maintaining reusable features and excellent pressure-holding capabilities up to 500 psi, making it ideal for sensitive analytical processes. Trust this superior construction to enhance performance and reliability in your lab applications.