Learn More
Waters Corp Waters Fingertight Fitting, PEEK, 6000 psi, 1/16 in. OD
Catalog No. 501385010 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PEEK One Piece Fingertight Fitting is designed for demanding applications, capable of withstanding pressures up to 6000 psi. Its single-piece nut and ferrule design, made from chemically inert PEEK material, ensures leak-tight seals. The fitting features a knurled head with an increased diameter for easy, tool-free tightening and is compatible with 1/16 in. OD tubing.
  • Withstands pressure up to 6000 psi (420 bar).
  • Made from a single piece of PEEK material for leak-tight seals.
  • Chemically inert materials prevent contamination.
  • Knurled head with increased diameter allows for tool-free tightening.
  • Compatible with 1/16 in. OD tubing.
Catalog No. 50-138-5010 Supplier Waters Corp Supplier No. 186008714
This PEEK One Piece Fingertight Fitting is designed for demanding applications, capable of withstanding pressures up to 6000 psi. Its single-piece nut and ferrule design, made from chemically inert PEEK material, ensures leak-tight seals. The fitting features a knurled head with an increased diameter for easy, tool-free tightening and is compatible with 1/16 in. OD tubing.
  • Withstands pressure up to 6000 psi (420 bar).
  • Made from a single piece of PEEK material for leak-tight seals.
  • Chemically inert materials prevent contamination.
  • Knurled head with increased diameter allows for tool-free tightening.
  • Compatible with 1/16 in. OD tubing.