Learn More
Waters Corp WATERS CORPORATION Syringe Adapter, PEEK Port, Rheodyne, ZQ Mass Detectors
Catalog No. 50792401 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation PEEK Port Rheodyne Syringe Adapter facilitates the connection of syringes to ZQ Mass Detectors. This adapter is also known as a PEEK Port or Rheodyne injector.
  • Used with external syringe pumps for ZQ 2000 systems.
  • Compatible with 500 μL syringes, such as the Hamilton model 1750.
  • Connects syringes to the divert/inject valve on the ZQ.
Catalog No. 50-792-401 Supplier Waters Corp Supplier No. 700000472
The Waters Corporation PEEK Port Rheodyne Syringe Adapter facilitates the connection of syringes to ZQ Mass Detectors. This adapter is also known as a PEEK Port or Rheodyne injector.
  • Used with external syringe pumps for ZQ 2000 systems.
  • Compatible with 500 μL syringes, such as the Hamilton model 1750.
  • Connects syringes to the divert/inject valve on the ZQ.