Learn More
Trajan Scientific PEEK TUBE PURPLE 1/32" OD X 0.012" ID, LENGTH 5 FEET
SDP
Catalog No. 502724814
Encompass_Preferred
PEEK TUBE PURPLE 1/32" OD X 0.012" ID, LENGTH 5 FEET
Catalog No. 50-272-4814 Supplier Trajan Scientific Supplier No. 13010120075F
PEEK TUBE PURPLE 1/32" OD X 0.012" ID, LENGTH 5 FEET