Learn More
Analytical Sales & Services PEEK Quick-Stop Luer Check Valve
Catalog No. 5000807079
Encompass
Quick-Stop Luer Check Valve\n• Prevents potentially hazardous spills\n• Place between solvent reservoir and pump\n• No pump re-priming with unique luer lock design\n\nQty/Pkg: EACH
Catalog No. 50-008-07079 Supplier Analytical Sales & Services Supplier No. 59697
Quick-Stop Luer Check Valve\n• Prevents potentially hazardous spills\n• Place between solvent reservoir and pump\n• No pump re-priming with unique luer lock design\n\nQty/Pkg: EACH