Learn More
Analytical Sales & Services PEEK Replacement Ferrule (Copy)
Catalog No. 5000804858
Encompass
PEEK Replacement Ferrule for 10/32 Fingertight for 1/16" OD Tubing\n\nQty/Pkg: EACH
Catalog No. 50-008-04858 Supplier Analytical Sales & Services Supplier No. 551421
PEEK Replacement Ferrule for 10/32 Fingertight for 1/16" OD Tubing\n\nQty/Pkg: EACH