Learn More
Analytical Sales & Services PEEK Replacement Needles for 250µL - 1mL Syringes
Catalog No. 5000806935
Encompass
PEEK Replacement Needles for 250µL - 1mL Syringes. 0.062 inch OD, 0.030 inch ID, 2" Length, Point Style #3\n\nQty/Pkg: 6PK
Catalog No. 50-008-06935 Supplier Analytical Sales & Services Supplier No. 48650014
PEEK Replacement Needles for 250µL - 1mL Syringes. 0.062 inch OD, 0.030 inch ID, 2" Length, Point Style #3\n\nQty/Pkg: 6PK