Learn More
Hach Company Hach PEEK Salt Bridge, Ceramic Outer-Junction for pHD Differential pH/ORP Sensors
Catalog No. NC2173621 Shop All Hach Company Products
Encompass
Replacement salt bridge for Hach pHD differential pH and ORP sensors. The PEEK body with ceramic outer junction is field-replaceable and is shipped in a salt solution to prevent dehydration; replace every 6 months for optimum performance.
  • PEEK body with ceramic outer junction
  • FKM/FPM O-ring for chemical compatibility
  • Shipped in salt solution to prevent dehydration
  • Field-replaceable double-junction design
  • Recommended replacement every 6 months for optimum performance
This item is not returnable. View return policy
Catalog No. NC2173621 Supplier Hach Company Supplier No. SBP2SV
Replacement salt bridge for Hach pHD differential pH and ORP sensors. The PEEK body with ceramic outer junction is field-replaceable and is shipped in a salt solution to prevent dehydration; replace every 6 months for optimum performance.
  • PEEK body with ceramic outer junction
  • FKM/FPM O-ring for chemical compatibility
  • Shipped in salt solution to prevent dehydration
  • Field-replaceable double-junction design
  • Recommended replacement every 6 months for optimum performance