Learn More
Analytical Sales & Services PEEK Short Flangeless Nut for 1/8" OD Tubing
Catalog No. 5000806882
Encompass
PEEK Short Flangeless Male Nut for 1/8" OD Tubing 1/4-28\n\nQty/Pkg: 10PK
Catalog No. 50-008-06882 Supplier Analytical Sales & Services Supplier No. 59335X
PEEK Short Flangeless Male Nut for 1/8" OD Tubing 1/4-28\n\nQty/Pkg: 10PK