Learn More
Analytical Sales & Services PEEK Super Flangeless Short Knurl Nut
Catalog No. 5000805084
Encompass
Super Flangeless Short Knurl, PEEK Nut, rated to 5000 psi, 10-32 Flat bottom port
Catalog No. 50-008-05084 Supplier Analytical Sales & Services Supplier No. 57653
Super Flangeless Short Knurl, PEEK Nut, rated to 5000 psi, 10-32 Flat bottom port