Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Tubing, PEEK, 1/16 in. x 0.020 in., 5 ft.

Supplier:  Waters Corp WAT022998

Encompass_Preferred

Waters™ tubing and fittings are designed to provide leak-free connections with low dead volume and minimal band spreading. These components are intended to be clean and work together, with matching geometry between the ferrule, conical port, and the diameter of the tube and fittings. A variety of materials, lengths, and diameters are available to offer flexibility for system setup and maintenance.

  • Designed for leak-free connections
  • Ensures low dead volume
  • Minimizes band spreading
  • Constructed from PEEK material
  • Features 1/16 in. outer diameter
  • Features 0.020 in. inner diameter
  • Provided in a 5 ft. length
  • Compatible with HPLC systems

Catalog No. 50-818-955


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.