Learn More
Waters Corp Waters Tubing, PEEK, 1/16 in x .007 in, 5 m
Catalog No. 50792292 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PEEK tubing is engineered for demanding laboratory applications, offering excellent chemical inertness and high-pressure resistance. Its smooth internal surface ensures consistent and reliable fluid transfer.
  • Constructed from PEEK material.
  • Features dimensions of 1/16 in x .007 in.
  • Supplied in a length of 5 m.
  • Identifiable by its yellow color.
Catalog No. 50-792-292 Supplier Waters Corp Supplier No. 6436004S5
This PEEK tubing is engineered for demanding laboratory applications, offering excellent chemical inertness and high-pressure resistance. Its smooth internal surface ensures consistent and reliable fluid transfer.
  • Constructed from PEEK material.
  • Features dimensions of 1/16 in x .007 in.
  • Supplied in a length of 5 m.
  • Identifiable by its yellow color.