Learn More
Waters Corp Waters Tubing, PEEK, 0.005 in. ID, 1/16 in. OD, 5 ft. Length
Catalog No. 50818954 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters™ tubing and fittings are designed to provide leak-free connections, with low dead volume, and minimal band spreading. For optimal performance, these components must be clean and designed to work together. The geometry of the ferrule and the conical port it is used with must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths, and diameters are available to provide flexibility in system setup and maintenance.
  • Designed to provide leak-free connections
  • Low dead volume
  • Minimal band spreading
  • Clean components for optimal performance
  • Optimized ferrule geometry for conical port connections
  • Matched tube and fitting diameters
  • Available in a variety of materials, lengths, and diameters for system setup and maintenance
  • Made from PEEK material
  • Suitable for HPLC systems
Catalog No. 50-818-954 Supplier Waters Corp Supplier No. WAT022995
Waters™ tubing and fittings are designed to provide leak-free connections, with low dead volume, and minimal band spreading. For optimal performance, these components must be clean and designed to work together. The geometry of the ferrule and the conical port it is used with must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths, and diameters are available to provide flexibility in system setup and maintenance.
  • Designed to provide leak-free connections
  • Low dead volume
  • Minimal band spreading
  • Clean components for optimal performance
  • Optimized ferrule geometry for conical port connections
  • Matched tube and fitting diameters
  • Available in a variety of materials, lengths, and diameters for system setup and maintenance
  • Made from PEEK material
  • Suitable for HPLC systems