Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .0025" ID x 100' Length
Catalog No. 5000804410
Encompass
PEEK Tubing - 1/16" OD x .0025" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 7,000psi (483 bar)
Catalog No. 50-008-04410 Supplier Analytical Sales & Services Supplier No. 41560XL4
PEEK Tubing - 1/16" OD x .0025" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 7,000psi (483 bar)