Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .007" ID x 100' Length
Catalog No. 5000804780
Encompass
PEEK Tubing - 1/16" OD x .007" ID x 100' Length\n\n\n\nColor: Yellow\n\n\n\nPressure Rating: 7,000psi (483 bar)
Catalog No. 50-008-04780 Supplier Analytical Sales & Services Supplier No. 41536XL4
Add to Cart
Add to Cart
PEEK Tubing - 1/16" OD x .007" ID x 100' Length\n\n\n\nColor: Yellow\n\n\n\nPressure Rating: 7,000psi (483 bar)