Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .010" ID x 100' Length
Catalog No. 5000806064
Encompass
PEEK Tubing - 1/16" OD x .010" ID x 100' Length\n\n\n\nColor: Blue\n\n\n\nPressure Rating: 7,000psi (483 bar)
Catalog No. 50-008-06064 Supplier Analytical Sales & Services Supplier No. 41531BXL4
PEEK Tubing - 1/16" OD x .010" ID x 100' Length\n\n\n\nColor: Blue\n\n\n\nPressure Rating: 7,000psi (483 bar)