Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .010" ID x 100' Length
Supplier:  Analytical Sales & Services 41531XL4
Encompass
PEEK Tubing - 1/16" OD x .010" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 7,000psi (483 bar)
Catalog No. 50-008-04817