Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .010" ID x 50' Length
Catalog No. 5000806412
Encompass
PEEK Tubing - 1/16" OD x .010" ID x 50' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 7,000psi (483 bar)
Catalog No. 50-008-06412 Supplier Analytical Sales & Services Supplier No. 41531L4
PEEK Tubing - 1/16" OD x .010" ID x 50' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 7,000psi (483 bar)