Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .020" ID x 100' Length
Catalog No. 5000806411
Encompass
PEEK Tubing - 1/16" OD x .020" ID x 100' Length\n\n\n\nColor: Orange\n\n\n\nPressure Rating: 6,000psi (414 bar)
Catalog No. 50-008-06411 Supplier Analytical Sales & Services Supplier No. 41532XL4
Add to Cart
Add to Cart
PEEK Tubing - 1/16" OD x .020" ID x 100' Length\n\n\n\nColor: Orange\n\n\n\nPressure Rating: 6,000psi (414 bar)