Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .020" ID x 50' Length
Catalog No. 5000806927
Encompass
PEEK Tubing - 1/16" OD x .020" ID x 50' Length\n\n\n\nColor: Orange\n\n\n\nPressure Rating: 6,000psi (414 bar)
Catalog No. 50-008-06927 Supplier Analytical Sales & Services Supplier No. 41532L4
PEEK Tubing - 1/16" OD x .020" ID x 50' Length\n\n\n\nColor: Orange\n\n\n\nPressure Rating: 6,000psi (414 bar)