Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .030" ID x 50' Length
Catalog No. 5000806928
Encompass
PEEK Tubing - 1/16" OD x .030" ID x 50' Length\n\n\n\nColor: Green\n\n\n\nPressure Rating: 4,000psi (276 bar)
Catalog No. 50-008-06928 Supplier Analytical Sales & Services Supplier No. 41533L4
Add to Cart
Add to Cart
PEEK Tubing - 1/16" OD x .030" ID x 50' Length\n\n\n\nColor: Green\n\n\n\nPressure Rating: 4,000psi (276 bar)