Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .040" ID x 100' Length
Catalog No. 5000806409
Encompass
PEEK Tubing - 1/16" OD x .040" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 3,000psi (207 bar)
Catalog No. 50-008-06409 Supplier Analytical Sales & Services Supplier No. 41538XL4
PEEK Tubing - 1/16" OD x .040" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 3,000psi (207 bar)