Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD x .055" ID x 100' Length
Catalog No. 5000806901
Encompass
PEEK Tubing - 1/16" OD x .055" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 500psi (34 bar)
Catalog No. 50-008-06901 Supplier Analytical Sales & Services Supplier No. 41537XL4
PEEK Tubing - 1/16" OD x .055" ID x 100' Length\n\n\n\nColor: Natural\n\n\n\nPressure Rating: 500psi (34 bar)