Learn More
Waters Corp Waters ACQUITY Tubing and Fittings Kit, PEEK
Catalog No. 502039483 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains various components, including fittings, PEEK screws, a PEEK tubing cutter, and different types of PEEK tubing, used to support various system configurations. It also includes a procedure document for configuring the UPLC System.
  • Includes multiple parts necessary for system configurations.
  • Contains a CAPLC PEEK tubing cutter for precise tubing length adjustment.
  • Comes with a procedure document for configuring the UPLC System, accessible via a QR Code on support.waters.com.
Catalog No. 50-203-9483 Supplier Waters Corp Supplier No. 700004067
This kit contains various components, including fittings, PEEK screws, a PEEK tubing cutter, and different types of PEEK tubing, used to support various system configurations. It also includes a procedure document for configuring the UPLC System.
  • Includes multiple parts necessary for system configurations.
  • Contains a CAPLC PEEK tubing cutter for precise tubing length adjustment.
  • Comes with a procedure document for configuring the UPLC System, accessible via a QR Code on support.waters.com.