Learn More
Microsolv Technology Corporation PEEK Tubing Drill 0.5mm. Stainless Steel Pin, Replacement for Flanging Tool. 1 EA.
Catalog No. 502271418
Encompass_Preferred
This Drill is designed to flange the tips of thin walled, PEEK tubing when used with the MicroSolv Flanging Tool.
This item is not returnable. View return policy
Catalog No. 50-227-1418 Supplier Microsolv Technology Corporation Supplier No. 45099105
This Drill is designed to flange the tips of thin walled, PEEK tubing when used with the MicroSolv Flanging Tool.