Learn More
Analytical Sales & Services PEEK Tubing - 1/16" OD X .040" ID X 50' Length
Catalog No. 50453748
Encompass
PEEK Tubing Natural, .040 x 1
Catalog No. 50-453-748 Supplier Analytical Sales & Services Supplier No. 41538L4
PEEK Tubing Natural, .040 x 1