Learn More
Shimadzu Scientific Instruments PEEK Tubing, Red, 0.062" OD x 42cm precut with white ID band
Catalog No. NC3313570
Encompass
Biocompatible, PEEK Tubing is optimized for use in high-pressure applications. Its distinct materials makes it inert to most solvents and gives it a smooth internal surface that improves the resolution of sample bands. This is a precision machined product for Shimadzu LCMS instruments. Laser cut and pre measured for use as the loop on the ESI probe for the LCMS-2020 and LCMS-8030/40/45/50/60. It has a white ID band to allow users to quickly identify the unique product. Color; Red with white ID band/label, Length (cm); 42cm +/- 0.2mm, Material; PEEK, Pressure Rating Max (psi); 7000, Pressure Rating Max (bar); 483, Tubing ID (in); 0.005, Tubing ID (mm); 0.125, Tubing OD (in); 1/16.
Catalog No. NC3313570 Supplier Shimadzu Scientific Instruments Supplier No. 2209124604
Biocompatible, PEEK Tubing is optimized for use in high-pressure applications. Its distinct materials makes it inert to most solvents and gives it a smooth internal surface that improves the resolution of sample bands. This is a precision machined product for Shimadzu LCMS instruments. Laser cut and pre measured for use as the loop on the ESI probe for the LCMS-2020 and LCMS-8030/40/45/50/60. It has a white ID band to allow users to quickly identify the unique product. Color; Red with white ID band/label, Length (cm); 42cm +/- 0.2mm, Material; PEEK, Pressure Rating Max (psi); 7000, Pressure Rating Max (bar); 483, Tubing ID (in); 0.005, Tubing ID (mm); 0.125, Tubing OD (in); 1/16.