Learn More
Waters Corp Waters Corporation Fingertight Fittings, PEEK, Two-Piece, 1/16 in., 10-32 Thread
Catalog No. 50784241 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The PEEK Ferrule, Fingertight Fittings are designed for ease of use with a 1/16 in. size and 10-32 thread. These fittings are made from PEEK material and are classified as Spare Parts Kits. They offer a reliable solution for various applications requiring secure, hand-tightened connections.
  • Fingertight fittings
  • Made of PEEK material
  • Pressure rating up to 5000 psi (345 bar)
  • Designed for 1/16-in. tubing
  • 10-32 thread type
Catalog No. 50-784-241 Supplier Waters Corp Supplier No. PSL613316
The PEEK Ferrule, Fingertight Fittings are designed for ease of use with a 1/16 in. size and 10-32 thread. These fittings are made from PEEK material and are classified as Spare Parts Kits. They offer a reliable solution for various applications requiring secure, hand-tightened connections.
  • Fingertight fittings
  • Made of PEEK material
  • Pressure rating up to 5000 psi (345 bar)
  • Designed for 1/16-in. tubing
  • 10-32 thread type