Learn More
Waters Corp Waters Corporation Union, PEEK, 2/pk, MS System
Catalog No. NC9667540 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PEEK union is a spare part designed for use as a fitting in MS systems. It is supplied in packages of two and can be replaced as required. It can also serve as a replacement for worn inlet/outlet ports on certain detectors.
  • Designed for MS systems
  • Made from PEEK material
  • Supplied as a pack of two
  • Functions as a fitting
  • Can be used for inlet/outlet port replacement
Catalog No. NC9667540 Supplier Waters Corp Supplier No. 415000106
This PEEK union is a spare part designed for use as a fitting in MS systems. It is supplied in packages of two and can be replaced as required. It can also serve as a replacement for worn inlet/outlet ports on certain detectors.
  • Designed for MS systems
  • Made from PEEK material
  • Supplied as a pack of two
  • Functions as a fitting
  • Can be used for inlet/outlet port replacement