Learn More
Thermo Scientific PEEK Viper Assembly Thermal Spray Coating 178mm
Catalog No. NC1711431 Shop All Thermo Scientific Products
Encompass
178 millimeter assembly made of polyether ether ketone engineering thermoplastic polymer with exceptional chemical and high temperature resistance, maintaining strength, stiffness and dimensional stability designed for demanding mechanical and electrical applications requiring durability and chemical inertness.
Catalog No. NC1711431 Supplier Thermo Scientific Supplier No. 088808
178 millimeter assembly made of polyether ether ketone engineering thermoplastic polymer with exceptional chemical and high temperature resistance, maintaining strength, stiffness and dimensional stability designed for demanding mechanical and electrical applications requiring durability and chemical inertness.