Learn More
IDEX Health & Science Upchurch PEEK Y ASSEMBLY 1/4-28 .020IN
Catalog No. NC1919882
Encompass_Preferred
Idex Low Pressure Y Connectors, Diameter Inner: 0.02 in., Includes: P-235/P-200 fittings, Swept Volume: 17 uL, PEEK, For Use With: Split a Stream or Join Two Streams Together, Pressure: 1000 psi
Catalog No. NC1919882 Supplier IDEX Health & Science Upchurch Supplier No. P512
Idex Low Pressure Y Connectors, Diameter Inner: 0.02 in., Includes: P-235/P-200 fittings, Swept Volume: 17 uL, PEEK, For Use With: Split a Stream or Join Two Streams Together, Pressure: 1000 psi