Learn More
IDEX Health & Science Upchurch PEEK Y ASSEMBLY 1/4-28 .060IN
Supplier:  IDEX Health & Science Upchurch P514
Encompass_Preferred
Idex Low Pressure Y Connectors, Diameter Inner: 0.06 in., Swept Volume: 13.6 uL, PEEK, For Use With: Split a Stream or Join Two Streams Together, Pressure: 1000 psi
Catalog No. NC1919883