Learn More
Analytical Sales & Services PEEKsil™ Tubing - 1/16" OD x 175µm ID, 150mm Length
Catalog No. 5000804711
Encompass
PEEKsil™ Tubing - 1/16" OD x 175µm ID, 150mm Length. Color: Yellow\n\n\n\n• Fused silica tubing covered in PEEK tubing\n\n• Do not cut this tubing\n\n• Use metal or PEEK fittings\n\n• Great alternative to PEEK tubing\n\n\n\nPressure Rating\n\n50 micron (0.002") ≤ 25,000 psi\n\n100 micron (0.004") ≤ 15,000 psi\n\n175 micron (0.007") ≤ 8,500 psi\n\n200 micron (0.008") ≤ 6,000 psi
Catalog No. 50-008-04711 Supplier Analytical Sales & Services Supplier No. 6175150
PEEKsil™ Tubing - 1/16" OD x 175µm ID, 150mm Length. Color: Yellow\n\n\n\n• Fused silica tubing covered in PEEK tubing\n\n• Do not cut this tubing\n\n• Use metal or PEEK fittings\n\n• Great alternative to PEEK tubing\n\n\n\nPressure Rating\n\n50 micron (0.002") ≤ 25,000 psi\n\n100 micron (0.004") ≤ 15,000 psi\n\n175 micron (0.007") ≤ 8,500 psi\n\n200 micron (0.008") ≤ 6,000 psi