Learn More
Analytical Sales & Services PEEKsil™ Tubing - 1/32" OD x 150µm ID, 500mm Length
Catalog No. 5000804823
Encompass
PEEKsil™ Tubing - 1/32" OD x 150µm ID, 500mm Length. Color: Purple\n\n\n\n• Fused silica tubing covered in PEEK tubing\n\n• Do not cut this tubing\n\n• Use metal or PEEK fittings\n\n• Great alternative to PEEK tubing\n\n\n\nPressure Rating\n\n50 micron (0.002") ≤ 25,000 psi\n\n100 micron (0.004") ≤ 15,000 psi\n\n175 micron (0.007") ≤ 8,500 psi\n\n200 micron (0.008") ≤ 6,000 psi
Catalog No. 50-008-04823 Supplier Analytical Sales & Services Supplier No. 3150500
PEEKsil™ Tubing - 1/32" OD x 150µm ID, 500mm Length. Color: Purple\n\n\n\n• Fused silica tubing covered in PEEK tubing\n\n• Do not cut this tubing\n\n• Use metal or PEEK fittings\n\n• Great alternative to PEEK tubing\n\n\n\nPressure Rating\n\n50 micron (0.002") ≤ 25,000 psi\n\n100 micron (0.004") ≤ 15,000 psi\n\n175 micron (0.007") ≤ 8,500 psi\n\n200 micron (0.008") ≤ 6,000 psi