Learn More
Polysciences, Inc. Mold, Peel-A-Way Embedding; Rectangular R-40; 40L x 22W x 20mm D; Clear plastic weighing tray; Embedding paraffin; Disposable embedding mold; Tissue capsule; Slide grip
SDP
Catalog No. NC9960096
Encompass_Preferred
Mold, Peel-A-Way Embedding; Rectangular R-40; 40L x 22W x 20mm D; Clear plastic weighing tray; Embedding paraffin; Disposable embedding mold; Tissue capsule; Slide grip
Catalog No. NC9960096 Supplier Polysciences, Inc. Supplier No. 18646C1
Mold, Peel-A-Way Embedding; Rectangular R-40; 40L x 22W x 20mm D; Clear plastic weighing tray; Embedding paraffin; Disposable embedding mold; Tissue capsule; Slide grip