Learn More
Polysciences, Inc. PEEL-A-WAY EMBEDDING MOLD polyethylene construction is designed to “Peel-A-Way” from the solidified block, eliminating the need for release agents and block trimming.
SDP
Catalog No. NC9574758
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
PEEL-A-WAY EMBEDDING MOLD SAMP
Catalog No. NC9574758 Supplier Polysciences, Inc. Supplier No. 18646D1
This item has a minimum qty of 2 per supplier requirements.
PEEL-A-WAY EMBEDDING MOLD SAMP