Learn More
Ted Pella Inc Peel-A-Way 22 x 40mm x 20mm deep, rectangular
SDP
Catalog No. NC0005138
Encompass
These tear away molds are useful for most epoxy and acrylic resin embedding media and for paraffin waxes. Especially suited for use with paraffin. These molds may be used under anaerobic conditions. Peel-A-Way I.D. Tabs are for use as identification with the Peel-A-Way embedding molds (8000 per book).
Catalog No. NC0005138 Supplier Ted Pella Inc Supplier No. 27114
These tear away molds are useful for most epoxy and acrylic resin embedding media and for paraffin waxes. Especially suited for use with paraffin. These molds may be used under anaerobic conditions. Peel-A-Way I.D. Tabs are for use as identification with the Peel-A-Way embedding molds (8000 per book).