Learn More
Polysciences, Inc. Microscope Slide Holder, Peel-A-Way, Each carton holds 6 of these convenient grips.
SDP
Catalog No. NC9871605
Encompass
Microscope Slide Holder, Peel-A-Way, Each carton holds 6 of these convenient grips. The holders are made of durable plastic, capable of withstanding temperatures up to 80deg.C as well as being resistant to chemicals and solvents
Catalog No. NC9871605 Supplier Polysciences, Inc. Supplier No. 19801A1
Microscope Slide Holder, Peel-A-Way, Each carton holds 6 of these convenient grips. The holders are made of durable plastic, capable of withstanding temperatures up to 80deg.C as well as being resistant to chemicals and solvents