Learn More
Electron Microscopy Sciences Peel Away Disposable Embedding Mold Truncated Shape, 12 x 12 mm
SDP
Catalog No. 501899235
Encompass_Preferred
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 12 x 12 mm
Catalog No. 50-189-9235 Supplier Electron Microscopy Sciences Supplier No. 70181
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 12 x 12 mm