Learn More
Electron Microscopy Sciences Peel Away Disposable Embedding Mold Rectangular, 22 x 30 mm
SDP
Catalog No. 501899236
Encompass_Preferred
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 22 x 30 mm
Catalog No. 50-189-9236 Supplier Electron Microscopy Sciences Supplier No. 70183
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 22 x 30 mm