Learn More
Bio Rad Laboratories PEELING FOIL HEAT SEAL 100/PK
Catalog No. NC0903289
Encompass
Pkg of 100, laminate/foil peelable seals for PCR and sample storage, for use with PX1 PCR plate sealer.
Catalog No. NC0903289 Supplier Bio Rad Laboratories Supplier No. 1814045
Pkg of 100, laminate/foil peelable seals for PCR and sample storage, for use with PX1 PCR plate sealer.