Learn More
MSC Peerless Chain Welded Chain, Alloy, Grade 80, 3/8 in, 20 ft
Catalog No. 5000527204
Encompass
This welded chain is an alloy chain designed for heavy-duty lifting applications. With a Grade 80 rating and a 3/8 inch trade size, it offers high strength and durability. The chain features a Thermadep finish and an impressive load capacity of 7100 lbs, making it suitable for various industrial uses.
  • Suitable for lifting applications.
  • Manufactured from alloy for durability.
  • Grade 80 for high strength.
  • 3/8 inch trade size.
  • Overall length of 20 feet.
  • Thermadep finish.
  • Load capacity of 7100 lbs.
  • Meets ASTM, OSHA, and NACM standards.
Catalog No. 50-005-27204 Supplier MSC Supplier No. 47473657
This welded chain is an alloy chain designed for heavy-duty lifting applications. With a Grade 80 rating and a 3/8 inch trade size, it offers high strength and durability. The chain features a Thermadep finish and an impressive load capacity of 7100 lbs, making it suitable for various industrial uses.
  • Suitable for lifting applications.
  • Manufactured from alloy for durability.
  • Grade 80 for high strength.
  • 3/8 inch trade size.
  • Overall length of 20 feet.
  • Thermadep finish.
  • Load capacity of 7100 lbs.
  • Meets ASTM, OSHA, and NACM standards.