Learn More
MSC Peerless Chain Welded Proof Coil Chain, 5/16 in, Grade 30, Carbon Steel
Catalog No. 5000530330
Encompass
This welded proof coil chain is constructed from carbon steel and features a self-colored finish. Designed for various applications, it has a trade size of 5/16 inch and a grade of 30. It is offered in 1-foot increments, with a total coil length of 550 feet.
  • 1,900 lb work load limit
  • Grade 30
  • Carbon steel material
  • Self-colored finish
  • Welded construction
  • 5/16 inch trade size
  • 0.4900 inch inside width
  • 1.1400 inch inside length
  • Manufactured in China
  • Cut-to-length items require special handling
Catalog No. 50-005-30330 Supplier MSC Supplier No. 67772749
This welded proof coil chain is constructed from carbon steel and features a self-colored finish. Designed for various applications, it has a trade size of 5/16 inch and a grade of 30. It is offered in 1-foot increments, with a total coil length of 550 feet.
  • 1,900 lb work load limit
  • Grade 30
  • Carbon steel material
  • Self-colored finish
  • Welded construction
  • 5/16 inch trade size
  • 0.4900 inch inside width
  • 1.1400 inch inside length
  • Manufactured in China
  • Cut-to-length items require special handling