Learn More
MSC Peerless Chain Welded Straight Link Machine Chain, #2, Steel, Standard Finish, 325 Lb Capacity
Catalog No. 5000527366
Encompass
This #2 Welded Straight Link Machine Chain from Peerless Chain is constructed from steel with a standard finish. It offers a work load limit of 325 Lb, making it suitable for various light-duty applications requiring reliable and durable chain.
  • Straight link machine chain design
  • Constructed from steel
  • Standard finish for general applications
  • Chain grade 30 for light-duty use
  • Work load limit of 325 Lb
  • Trade size #2 for specific applications
  • Inside width of 0.2600 in
  • Inside length of 0.6100 in
  • Manufactured in the United States
Catalog No. 50-005-27366 Supplier MSC Supplier No. 67773861
This #2 Welded Straight Link Machine Chain from Peerless Chain is constructed from steel with a standard finish. It offers a work load limit of 325 Lb, making it suitable for various light-duty applications requiring reliable and durable chain.
  • Straight link machine chain design
  • Constructed from steel
  • Standard finish for general applications
  • Chain grade 30 for light-duty use
  • Work load limit of 325 Lb
  • Trade size #2 for specific applications
  • Inside width of 0.2600 in
  • Inside length of 0.6100 in
  • Manufactured in the United States