Learn More
MSC Peerless Chain Welded Transport Chain, 1/4 in, Grade 70, Carbon Steel
Catalog No. 5000516037
Encompass
This welded transport chain offers robust performance for demanding applications. Constructed from durable carbon steel with a yellow chromate zinc finish, it provides excellent resistance to wear and corrosion. Rated at Grade 70 with a substantial 3,150 Lb capacity, this 1/4 inch chain is suitable for securing heavy loads during transport or in industrial settings. It is available in 1' increments, with a total coil length of 800 ft.
  • 3,150 Lb capacity
  • Grade 70 for heavy-duty applications
  • Durable carbon steel construction
  • Yellow chromate zinc finish for corrosion resistance
  • 1/4 in trade size
  • 800 ft total coil length
  • Inside width of 0.45 in
  • Inside length of 1.24 in
Catalog No. 50-005-16037 Supplier MSC Supplier No. 67772285
This welded transport chain offers robust performance for demanding applications. Constructed from durable carbon steel with a yellow chromate zinc finish, it provides excellent resistance to wear and corrosion. Rated at Grade 70 with a substantial 3,150 Lb capacity, this 1/4 inch chain is suitable for securing heavy loads during transport or in industrial settings. It is available in 1' increments, with a total coil length of 800 ft.
  • 3,150 Lb capacity
  • Grade 70 for heavy-duty applications
  • Durable carbon steel construction
  • Yellow chromate zinc finish for corrosion resistance
  • 1/4 in trade size
  • 800 ft total coil length
  • Inside width of 0.45 in
  • Inside length of 1.24 in